Learn More
Abnova™ IRX2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00153572T01
Catalog No. 89-949-219
Add to cart
Specifications
Specifications
293T
NM_033267.2
pCMV-IRX2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
IRX2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 51.92kDa
Transient overexpression cell lysate was tested with anti-IRX2 antibody (Mfr. No. H00153572-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human