Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRX6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179257

 View more versions of this product

Catalog No. NBP179257


Add to cart

Description

Description

IRX6 Polyclonal antibody specifically detects IRX6 in Human, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

IRX6
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
IRX6
Synthetic peptide directed towards the N terminal of human IRX6The immunogen for this antibody is IRX6. Peptide sequence SFPHFGHPYRGASQFLASASSSTTCCESTQRSVSDVASGSTPAPALCCAP.
Immunogen affinity purified
RUO
Primary
79190
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_077311
iroquois homeobox 6, Iroquois homeobox protein 6Homeodomain protein IRXB3, iroquois homeobox protein 7, iroquois-class homeodomain protein IRX-6, IRX-3, IRX7IRXB3
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%; Bovine: 85%; Canine: 85%; Guinea pig: 85%; Equine: 85%; Pig: 85%; Rat: 84%.
Bovine, Canine, Equine, Guinea Pig, Human, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit