Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRX6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | IRX6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179259
|
Novus Biologicals
NBP179259 |
100 μL |
Each of 1 for $436.00
|
|
Description
IRX6 Polyclonal specifically detects IRX6 in Human samples. It is validated for Western Blot.Specifications
IRX6 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
iroquois homeobox 6, Iroquois homeobox protein 6Homeodomain protein IRXB3, iroquois homeobox protein 7, iroquois-class homeodomain protein IRX-6, IRX-3, IRX7IRXB3 | |
IRX6 | |
IgG | |
Affinity Purified | |
48 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_077311 | |
79190 | |
Synthetic peptide directed towards the N terminal of human IRX6The immunogen for this antibody is IRX6. Peptide sequence SDVASGSTPAPALCCAPYDSRLLGSARPELGAALGIYGAPYAAAAAAQSY. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title