Learn More
Tocris Bioscience™ Isamoltane hemifumarate Available on GSA/VA Contract for Federal Government customers only.
5-HT1B antagonist
Manufacturer:  Tocris Bioscience™ 0992/10
 View more versions of this product
Catalog No. 09-921-0
Add to cart
Description
Description
5-HT1B antagonist, approximately 30-fold selective over 5-HT1A. Also possesses affinity for β-adrenergic receptors.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1-[(1-Methylethyl)amino]-3-[2-(1H-pyrrol-1-yl)]-propan-2-ol hemifumarate
>99%
Observed MW: 332.4
Soluble to 10mM with gentle warming in water
874882-92-5
C16H22N2O2.½C4H4O4
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at +4°C