Learn More
Tocris Bioscience™ Isamoltane hemifumarate
5-HT1B antagonist
Manufacturer:  Tocris Bioscience™ 0992/50
 View more versions of this product
Catalog No. 09-925-0
Add to cart
Description
Description
5-HT1B antagonist, approximately 30-fold selective over 5-HT1A. Also possesses affinity for β-adrenergic receptors.
Specifications
Specifications
1-[(1-Methylethyl)amino]-3-[2-(1H-pyrrol-1-yl)]-propan-2-ol hemifumarate
>99%
Observed MW: 332.4
Soluble to 10mM with gentle warming in water
874882-92-5
C16H22N2O2.½C4H4O4
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at +4°C