Alfa Aesar™ Isatin-3-oxime, 97%

Manufacturer: Alfa Aesar™ B2273206

Quantity 5g

 View more versions of this product

Catalog No. AAB2273206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 607-28-3
    Molecular Formula C8H6N2O2
    Formula Weight 162.15
    MDL Number MFCD00014568
    Beilstein 136198
    Synonym beta-Isatoxime
    Chemical Name or Material Isatin-3-oxime

    Specifications

    Formula Weight 162.15
    Molecular Formula C8H6N2O2
    MDL Number MFCD00014568
    CAS 607-28-3
    Beilstein 136198
    Synonym beta-Isatoxime
    Melting Point ≈228°C (dec.)
    RTECS Number NL7980000
    Quantity 5g
    Chemical Name or Material Isatin-3-oxime
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit