Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isatin-3-oxime, 97%

Click to view available options
Quantity:
5g
25g
100g
607-28-3
C8H6N2O2
162.148
MFCD00014568
LNMAXZZQNSPQSR-UHFFFAOYSA-N
isatin-3-oxime,3-hydroxyimino indolin-2-one,1h-indole-2,3-dione 3-oxime,3-oximeindole-2,3-dione,isatin beta-oxime,1h-indole-2,3-dione, 3-oxime,3-hydroxyiminoindolin-2-one,indole-2,3-dione, 3-oxime,3e-1h-indole-2,3-dione 3-oxime,3-hydroxyamino indol-2-one
5351629
3-(hydroxyamino)indol-2-one
C1=CC2=C(C(=O)N=C2C=C1)NO
607-28-3
C8H6N2O2
162.148
MFCD00014568
LNMAXZZQNSPQSR-UHFFFAOYSA-N
isatin-3-oxime,3-hydroxyimino indolin-2-one,1h-indole-2,3-dione 3-oxime,3-oximeindole-2,3-dione,isatin beta-oxime,1h-indole-2,3-dione, 3-oxime,3-hydroxyiminoindolin-2-one,indole-2,3-dione, 3-oxime,3e-1h-indole-2,3-dione 3-oxime,3-hydroxyamino indol-2-one
5351629
3-(hydroxyamino)indol-2-one
C1=CC2=C(C(=O)N=C2C=C1)NO
Melting Point ~228°C (decomposition)
Quantity 100g
Beilstein 136198
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Isatin-3-oxime

RTECSNumber : NL7980000