Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isatin 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
91-56-5
C8H5NO2
147.13
MFCD00005718
JXDYKVIHCLTXOP-UHFFFAOYSA-N
isatin,indoline-2,3-dione,2,3-indolinedione,indole-2,3-dione,isatine,2,3-dioxoindoline,pseudoisatin,tribulin,isotin,isatic acid lactam
7054
CHEBI:27539
1H-indole-2,3-dione
C1=CC=C2C(=C1)C(=O)C(=O)N2
91-56-5
C8H5NO2
147.13
MFCD00005718
JXDYKVIHCLTXOP-UHFFFAOYSA-N
isatin,indoline-2,3-dione,2,3-indolinedione,indole-2,3-dione,isatine,2,3-dioxoindoline,pseudoisatin,tribulin,isotin,isatic acid lactam
7054
CHEBI:27539
1H-indole-2,3-dione
C1=CC=C2C(=C1)C(=O)C(=O)N2
Flash Point 220°C
Color Orange
Infrared Spectrum Authentic
Quantity 500g
Melting Point 201°C to 204°C
Packaging Glass bottle
Sulfated Ash 0.1% max.
Assay Percent Range 98%
Beilstein 21, 432
Merck Index 15, 5148
Solubility Information Solubility in water: 1.9g/L. Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,diethyl ether,benzene
Formula Weight 147.13
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Isatin, 98%
Show More Show Less

EINECSNumber : 202-077-8

RTECSNumber : NL7873000

TSCA : TSCA