Learn More
Aviva Systems Biology Corp ISCA2 Antibody - middle region (ARP53448_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ISCA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ISCA2. Alternate names: HBLD1, ISA2, c14_5557ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61027