Learn More
iSEC1 Rat anti-Mouse,Rat, Unconjugated, Clone: GT42, MilliporeSigma™
Rat Monoclonal Antibody
Manufacturer:  MilliporeSigma MABF2226100UG
 View more versions of this product
Catalog No. MABF22261MI
Add to cart
Description
Description
iSEC1 Monoclonal antibody specifically detects iSEC1 in Mouse,Rat samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
Specifications
iSEC1
GT42
Purified
Gm609, Isec1
iSEC1-expressing NIH/3T3 cells.
34.68 kDa calculated.
100 μg
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal
Mouse, Rat
Flow Cytometry, Immunocytochemistry
Unconjugated
Intestinal secretory protein 1
Rat
IgG1κ
Protein G Purified
Inflammation and Immunology
Primary
GT42 is a rat monoclonal antibody that specifically detects murine ISEC1 (intestinal secretory cell protein 1), but not the iSEC2.