Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iSEC1 Rat anti-Mouse,Rat, Unconjugated, Clone: GT42, MilliporeSigma™

Rat Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen iSEC1
Applications Flow Cytometry, Immunocytochemistry
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22261MI
View Documents
MilliporeSigma
MABF2226100UG
100 μg
Each for $386.00
Add to cart
 
MABF22262MI
View Documents
MilliporeSigma
MABF222625UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

iSEC1 Monoclonal antibody specifically detects iSEC1 in Mouse,Rat samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications

Specifications

iSEC1
Unconjugated
Rat
Intestinal secretory protein 1
iSEC1-expressing NIH/3T3 cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry, Immunocytochemistry
Purified
Mouse, Rat
Gm609, Isec1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit