Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isethionic acid, sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
1562-00-1
C2H5NaO4S
148.11
MFCD00007534
LADXKQRVAFSPTR-UHFFFAOYSA-M
sodium 2-hydroxyethanesulfonate,sodium isethionate,isethionic acid sodium salt,2-hydroxyethanesulfonic acid sodium salt,sodium hydroxyethylsulfonate,ethanesulfonic acid, 2-hydroxy-, monosodium salt,sodium 2-hydroxyethylsulfonate,sodium 2-hydroxyethanesulphonate,sodium beta-hydroxyethanesulfonate,sodium 1-hydroxy-2-ethanesulfonate
517063
sodium;2-hydroxyethanesulfonate
C(CS(=O)(=O)[O-])O.[Na+]
1562-00-1
C2H5NaO4S
148.11
MFCD00007534
LADXKQRVAFSPTR-UHFFFAOYSA-M
sodium 2-hydroxyethanesulfonate,sodium isethionate,isethionic acid sodium salt,2-hydroxyethanesulfonic acid sodium salt,sodium hydroxyethylsulfonate,ethanesulfonic acid, 2-hydroxy-, monosodium salt,sodium 2-hydroxyethylsulfonate,sodium 2-hydroxyethanesulphonate,sodium beta-hydroxyethanesulfonate,sodium 1-hydroxy-2-ethanesulfonate
517063
sodium;2-hydroxyethanesulfonate
C(CS(=O)(=O)[O-])O.[Na+]
Water 1.5% max.
Color White
Melting Point 191.0°C to 194.0°C
Packaging Plastic bottle
Quantity 1kg
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula HOCH2CH2SO3Na
Beilstein 04, III, 42
Merck Index 14, 5107
Solubility Information Solubility in water: soluble in water
Formula Weight 148.11
Physical Form Fine Powder
Percent Purity 98%
Chemical Name or Material Isethionic acid, sodium salt, 98%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 216-343-6

TSCA : TSCA