Learn More
Abnova™ ISG20L1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00064782T01
Catalog No. 89-949-232
Add to cart
Description
Description
Specifications
Specifications
293T
NM_022767.2
pCMV-ISG20L1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
ISG20L1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 35.86kDa
Transient overexpression cell lysate was tested with anti-ISG20L1 antibody (Mfr. No. H00064782-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human