Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ISG20L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104292
Description
Antibody detects endogenous levels of total IL20L2.
3'-> 5'-exoribonuclease involved in ribosome biogenesis in the processing of the 12S pre-rRNA. Displays a strong specificity for a 3'-end containing a free hydroxyl group.
Specifications
ISG20L2 | |
Polyclonal | |
Unconjugated | |
ISG20L2 | |
HSD38; HSD-38; interferon stimulated exonuclease gene 20 like 2; interferon stimulated exonuclease gene 20kDa like 2; interferon stimulated exonuclease gene 20kDa-like 2; interferon-stimulated 20 kDa exonuclease-like 2; ISG20L2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
81875 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H9L3 | |
ISG20L2 | |
A synthesized peptide derived from human ISG20L2(Accession Q9H9L3), corresponding to amino acid residues I129-M179. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction