Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Islet-1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271050AF405

Catalog No. N271050A405


Add to cart

Description

Description

Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Islet-1
OTI2D8
Western Blot
Affinity Purified, Conjugated
ISL1
Recombinant protein fragment corresponding to amino acids 134-349 of human Islet-1 (NP_002193) produced in E.coli.
Immunogen affinity purified
Diabetes Research, Lipid and Metabolism, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
3670.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit