Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ismatec™ Pump Tubing, 2-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone

Cole-Parmer™ Ismatec™ Pump Tubing, 2-Stop Color-Coded Autoanalysis/Microbore is designed for optimal operation in Ismatec pumps

Specifications

Autoclavable No
Color Opaque
For Use With (Application) Ismatec pumps
Material Platinum-cured Silicone
Type 2-stop Tubing
View More Specs

Products 6
Catalog Number Tubing Size Price Quantity & Availability  
Catalog Number Tubing Size Price Quantity & Availability  
13-200-053
View Documents
0.89 mm ID x 24 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-054
View Documents
1.02 mm ID x 31 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-055
View Documents
1.14 mm ID x 38 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-056
View Documents
1.42 mm ID x 55 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-057
View Documents
2.06 mm ID x 100 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-058
View Documents
2.79 mm ID x 180 mm L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • No leachable additives or plasticizers
  • Appearance: Opaque
  • Temperature: -70 to 52°C
  • Life at 100 rpm: 1000 hrs
Specifications

Specifications

No
Ismatec pumps
2-stop Tubing
Opaque
Platinum-cured Silicone
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit