Learn More
Fisherbrand™ ISO 5&6 Cleanroom Stainless Steel Table with Casters and Perforated Top
Fixed Height Cleanroom Stainless Steel Tables with Casters & Perforated Top have a heavy duty all Type 304 stainless steel C-frame construction.
Specifications
Adjustable No
Adjustable (Height) No
Autoclavable Not Autoclavable
Certifications/Compliance SEFA 8
Depth (English) 30 in.
View More Specs
Products 3
Catalog Number Width (English) Price Quantity & Availability  
Catalog Number Width (English) Price Quantity & Availability  
15-050-290
View Documents
48 in. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
15-050-291
View Documents
60 in. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
15-050-292
View Documents
72 in. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description
Description
  • Fixed Height Stainless Steel Tables with Casters have a heavy-duty stainless-steel C-frame construction; suitable for ISO 5&6 Cleanroom environments
  • The perforated work top is Type 304 stainless steel, has a brushed finish with rolled edges at the front and back, and square turndown sides providing for flush adjacent installations when required.
  • These tables are furnished at a standing height with stainless steel legs that have adjustable bullet style 1 in. wide leveling feet.
  • Tables ship knocked down.
Specifications
Specifications
No
Not Autoclavable
30 in.
#4 Finish
36.125 in.
Table Assembly
None
2 Years
Yes
No
SEFA 8
76.2 cm
Stainless Steel
91.8 cm
None
None
Standard
Silver