Learn More
Fisherbrand™ ISO 5&6 Cleanroom Stainless Steel Table with Casters and Solid Top
Fixed Height Cleanroom Stainless Steel Tables with Casters have a heavy duty all Type 304 stainless steel C-frame construction.
 View more versions of this product
Catalog No. 15-050-284
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Fixed Height Stainless Steel Tables with Casters have a heavy-duty stainless-steel C-frame construction; suitable for ISO 5 & 6 Cleanroom environments
  • The work top is Type 304 stainless steel, has a brushed finish with rolled edges at the front and back, and square turndown sides providing for flush adjacent installations when required.
  • These tables are furnished at a standing height with stainless steel legs that have adjustable bullet style 1 in. wide leveling feet.
  • Optional Type 304 stainless steel undershelf.
  • Tables ship knocked down.
Specifications
Specifications
Stainless Steel
Yes
Standard
Not Autoclavable
No
No
48 in.
2 Years
36.125 in.
#4 Finish
SEFA 8
Table Assembly
Silver
None
None
None
91.8 cm
76.2 cm
30 in.
121.9 cm
Recommendations
Recommendations