Learn More
Fisherbrand™ ISO 5&6 Cleanroom Stainless Steel Table with Leveling Glides and Solid Top
Fixed Height Cleanroom Stainless Steel Tables with Leveling Glides have a heavy duty all Type 304 stainless steel C-frame construction.
 View more versions of this product
Catalog No. 15-050-280
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Fixed Height Stainless Steel Tables with Leveling Glides have a heavy-duty stainless-steel C-frame construction; suitable for ISO 5 & 6 Cleanroom environments
  • The work top is Type 304 stainless steel, has a brushed finish with rolled edges at the front and back, and square turndown sides providing for flush adjacent installations when required.
  • These tables are furnished at a standing height with stainless steel legs that have adjustable bullet style 1 in. wide leveling feet.
  • Optional Type 304 stainless steel undershelf.
  • Tables ship knocked down.
Specifications
Specifications
No
30 in.
2 Years
72 in.
#4 Finish
Table Assembly
Stainless Steel
None
None
No
91.8 cm
76.2 cm
36.125 in.
182.9 cm
Silver
SEFA 8
Not Autoclavable
None
No
Recommendations
Recommendations