Learn More
Fisherbrand™ ISO 7 & 8 Fixed Height Cleanroom Stainless Steel Table with Leveling Glides and Solid Top PROMO
ISO 7 & 8 Cleanroom Stainless Steel Table with Leveling Glides and Solid Top is a Fixed Height Cleanroom Table with Leveling Glides and a heavy duty all Type 304 stainless steel C-frame construction.
 View more versions of this product
Catalog No. 15-050-261PM
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Fixed Height Stainless Steel Tables with Leveling Glides have a heavy-duty stainless-steel C-frame construction.
  • Suitable for ISO 7 & 8 Cleanroom environments.
  • The work top is Type 304 stainless steel, has a brushed finish with rolled edges at the front and back, and square turndown sides, providing for flush adjacent installations when required.
  • These tables are furnished at a standing height with stainless steel legs that have adjustable bullet style 1 in. wide leveling feet.
  • Optional Type 304 stainless steel height adjustable undershelf.
  • Tables ship knocked down.
Specifications
Specifications
None
No
No. 4 Finish
76.2 cm
2 Years
30 in.
Stainless Steel
152.4 cm
Not Autoclavable
None
No
No
91.8 cm
36.125 in.
60 in.
Silver
Table Assembly
SEFA 8
None
Recommendations
Recommendations