Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%, Honeywell Burdick & Jackson

Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
540-84-1
C8H18
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
540-84-1
C8H18
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
CAS Max % 100.0000%
UN Number UN1262
Beilstein 1696876
Solubility Information 0.02% in Water at 20°C
Formula Weight 114.23
Percent Purity >99%
Grade ACS Reagent
Density 0.69 g/cm3
Boiling Point 99.24°C
Quantity 19 L
Flash Point −8°C
Packaging Metal Can
Water 0.01% max.
Assay by GC Analysis
Chemical Name or Material Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%
Show More Show Less

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes