Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
107-85-7
C5H13N
87.15
MFCD00008203
BMFVGAAISNGQNM-UHFFFAOYSA-N
isoamylamine,isopentylamine,3-methylbutylamine,1-amino-3-methylbutane,monoisoamylamine,leucamine,1-butanamine, 3-methyl,3-methylbutanamine,isovalerylamine,monoisopentylamine
7894
CHEBI:43689
3-methylbutan-1-amine
CC(C)CCN
107-85-7
C5H13N
87.15
MFCD00008203
BMFVGAAISNGQNM-UHFFFAOYSA-N
isoamylamine,isopentylamine,3-methylbutylamine,1-amino-3-methylbutane,monoisoamylamine,leucamine,1-butanamine, 3-methyl,3-methylbutanamine,isovalerylamine,monoisopentylamine
7894
CHEBI:43689
3-methylbutan-1-amine
CC(C)CCN
Viscosity 0.6 mPa.s (20°C)
Packaging Glass bottle
Color Colorless
Quantity 25mL
Flash Point 4°C
Boiling Point 95.0°C to 97.0°C
Melting Point -60.0°C
Refractive Index 1.4070 to 1.4100
Water 0.5% max.
Specific Gravity 0.75
Infrared Spectrum Authentic
Density 0.7500g/mL
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3)2CHCH2CH2NH2
UN Number 1106
Beilstein 04, 180
Merck Index 15, 5157
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
Formula Weight 87.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Isoamylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-526-