Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isobutanesulfonyl chloride, 97%

Click to view available options
Quantity:
250mg
1g
5g
35432-36-1
C4H9ClO2S
156.624
MFCD07369800
NQLNHTOCFWUYQE-UHFFFAOYSA-N
isobutanesulfonyl chloride,isobutylsulfonyl chloride,1-propanesulfonyl chloride, 2-methyl,2-methylpropane-1-sulfonylchloride,2-methyl-1-propylsulfonyl chloride,2-methyl-1-propanesulfonyl chloride,2-methyl-propane-1-sulfonyl chloride,isobutane sulfonyl chloride,isobutane-1-sulfonylchloride,acmc-1ae06
182236
2-methylpropane-1-sulfonyl chloride
CC(C)CS(=O)(=O)Cl
35432-36-1
C4H9ClO2S
156.624
MFCD07369800
NQLNHTOCFWUYQE-UHFFFAOYSA-N
isobutanesulfonyl chloride,isobutylsulfonyl chloride,1-propanesulfonyl chloride, 2-methyl,2-methylpropane-1-sulfonylchloride,2-methyl-1-propylsulfonyl chloride,2-methyl-1-propanesulfonyl chloride,2-methyl-propane-1-sulfonyl chloride,isobutane sulfonyl chloride,isobutane-1-sulfonylchloride,acmc-1ae06
182236
2-methylpropane-1-sulfonyl chloride
CC(C)CS(=O)(=O)Cl
Quantity 250mg
Color Colorless
Sensitivity Moisture Sensitive
Density 1.20g/mL
Refractive Index 1.4540
Linear Formula (CH3)2CHCH2SO2Cl
UN Number UN3265
Beilstein 1748728
Formula Weight 156.62
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Isobutanesulfonyl chloride
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.