Isobutanol, ACS, 99%, Alfa Aesar

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.001%, Solubility in water P.T., Titratable acid 0.0005meq/g, H2O 0.1%, Carbonyl compounds 0.01% (butyraldehyde) and 0.02% (2-butanone)

Manufacturer: Alfa Aesar 36643M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA36643M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-83-1
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C4H10O
    Formula Weight 74.12
    MDL Number MFCD00004740
    Synonym Isobutyl alcohol
    Chemical Name or Material Isobutanol
    Grade ACS Grade

    Specifications

    Physical Form Liquid
    Formula Weight 74.12
    Color Clear colorless
    Quantity 4 x 1L
    DOT Information Hazard Class: 3, Packing Group: III
    EINECS Number 201-148-0
    Odor Sweet
    Assay Percent Range >99%
    Molecular Formula C4H10O
    MDL Number MFCD00004740
    CAS 78-83-1
    Ignition Point 390°C (734°F)
    Synonym Isobutyl alcohol
    Health Hazard 1 H318-H315-H226-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness.
    Vapor Pressure 4-40 hPa (3-30mm Hg) at 20°C
    Appearance Colorless liquid
    Chemical Name or Material Isobutanol, For analysis ACS
    Grade ACS Grade

    H318-H315-H226-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit