Isobutanol, Spectrophotometric Grade, >99%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.10%

Manufacturer: Alfa Aesar 32433K2

Quantity 1L

 View more versions of this product

Catalog No. AA32433K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-83-1
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C4H10O
    Formula Weight 74.12
    MDL Number MFCD00004740
    Chemical Name or Material Isobutanol
    Grade Spectrophotometric grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 74.12
    Quantity 1L
    DOT Information Hazard Class: 3, Packing Group: III
    Density 0.802g/cm3
    EINECS Number 201-148-0
    Ignition Point 390°C (734°F)
    Odor Sweet
    Vapor Pressure 4-40 hPa (3-30mm Hg) at 20°C
    Assay Percent Range >99%
    Molecular Formula C4H10O
    MDL Number MFCD00004740
    CAS 78-83-1
    Health Hazard 1 H318-H315-H226-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness.
    Chemical Name or Material Isobutanol
    Grade Spectrophotometric grade

    H318-H315-H226-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit