Isobutyl Alcohol (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 1L

 View more versions of this product

Catalog No. A397-1

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 78-83-1
    Assay Percent Range ≥99.0 %
    Molecular Formula C4H10O
    Linear Formula (CH3)2CHCH2OH
    Formula Weight 74.12
    MDL Number MFCD00004740
    Synonym 1-Hydroxymethylpropane
    Chemical Name or Material Isobutyl Alcohol
    Assay ≥99.0%
    Grade Certified ACS Grade

    Specifications

    Formula Weight 74.12
    Boiling Point 108°C
    Melting Point −108°C
    Quantity 1L
    UN Number 1120
    Flash Point 28°C
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Colorless
    Density 2.6 (Air = 1.0)
    Vapor Pressure 11.7mbar at 20°C
    Molecular Formula C4H10O
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Irritating to respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Seek immediate medical attention/advice. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

    NFPA

    Health:2
    Flammability:3
    Instability:0
    Assay Percent Range ≥99.0 %
    Identification Pass Test
    Synonym 1-Hydroxymethylpropane
    Packaging Glass Bottle
    Residue after Evaporation 0.001% max.
    Additional Information Vapor Pressure: 11.7mbar at 20°C
    Solubility in Water Pass Test
    Butyraldehyde 0.01% max.
    2_Butanone 0.02% max.
    Cleanup Symbol Solvent
    Fluorescence Background Pass Test
    MDL Number MFCD00004740
    ChemAlert Storage Symbol Red
    Health Hazard 2 DANGER!
    Water 0.1% max.
    Titratable Acid 0.0005mEq/g max.
    Linear Formula (CH3)2CHCH2OH
    CAS 78-83-1
    Chemical Name or Material Isobutyl Alcohol
    Assay ≥99.0%
    Grade Certified ACS Grade
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. Irritating to respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Seek immediate medical attention/advice. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

    NFPA

    Health:2
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit