Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl benzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
120-50-3
C11H14O2
178.231
MFCD00048344
KYZHGEFMXZOSJN-UHFFFAOYSA-N
isobutyl benzoate,benzoic acid, isobutyl ester,benzoic acid, 2-methylpropyl ester,unii-kq6xz9wjii,isobutyl benzoate natural,fema no. 2185,kq6xz9wjii,benzoic acid isobutyl ester,benzoic acid, isobutyl ester 6ci,7ci,8ci,iso-butyl benzoate
61048
CHEBI:87500
2-methylpropyl benzoate
CC(C)COC(=O)C1=CC=CC=C1
120-50-3
C11H14O2
178.231
MFCD00048344
KYZHGEFMXZOSJN-UHFFFAOYSA-N
isobutyl benzoate,benzoic acid, isobutyl ester,benzoic acid, 2-methylpropyl ester,unii-kq6xz9wjii,isobutyl benzoate natural,fema no. 2185,kq6xz9wjii,benzoic acid isobutyl ester,benzoic acid, isobutyl ester 6ci,7ci,8ci,iso-butyl benzoate
61048
CHEBI:87500
2-methylpropyl benzoate
CC(C)COC(=O)C1=CC=CC=C1
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Quantity 25g
Color Colorless
Density 0.999g/mL
Assay Percent Range 98%
Linear Formula C6H5C(O)OCHCH(CH3)
Formula Weight 178.23
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Isobutyl benzoate, 98%
Show More Show Less

EINECSNumber : 204-401-3

RTECSNumber : DH4185000

TSCA : TSCA