Alfa Aesar™ Isobutyl benzoate, 99%

Manufacturer: Alfa Aesar™ A101140B

Quantity 1000g

 View more versions of this product

Catalog No. AAA101140B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 120-50-3
    Molecular Formula C11H14O2
    Formula Weight 178.23
    MDL Number MFCD00048344
    Beilstein 2045961
    Synonym Benzoic acid isobutyl ester
    Chemical Name or Material Isobutyl benzoate

    Specifications

    Formula Weight 178.23
    TSCA TSCA
    Boiling Point 242°C
    Flash Point 96°C (204°F)
    Molecular Formula C11H14O2
    MDL Number MFCD00048344
    CAS 120-50-3
    Beilstein 2045961
    Synonym Benzoic acid isobutyl ester
    RTECS Number DH4185000
    Quantity 1000g
    Chemical Name or Material Isobutyl benzoate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit