Isobutyl chloroformate, 98%

Manufacturer: Alfa Aesar A1469214

Quantity 25g

 View more versions of this product

Catalog No. AAA1469214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 543-27-1
    Molecular Formula ClCO2CH2CH(CH3)2
    Linear Formula CO2CH2CH(CH3)2
    Formula Weight 136.58
    MDL Number MFCD00000642
    UN Number UN2742
    Beilstein 956590
    Merck Index 14,5139
    Chemical Name or Material Isobutyl Chloroformate

    Specifications

    Formula Weight 136.58
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    Linear Formula CO2CH2CH(CH3)2
    MDL Number MFCD00000642
    UN Number UN2742
    Beilstein 956590
    Merck Index 14,5139
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318-H226
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.
    Physical Form Liquid
    Density 1.048
    Assay Percent Range 98%
    Molecular Formula ClCO2CH2CH(CH3)2
    CAS 543-27-1
    Storage Note 1 Moisture sensitive
    Chemical Name or Material Isobutyl Chloroformate
    Assay 98%

    Description

    Applications
    Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

    Solubility
    Immiscible with water.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.

    GHS H Statement
    H300-H310-H330-H314-H318-H226
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

    Moisture sensitive

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit