Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isobutyl dodecanoate, 99+%

Manufacturer:  Alfa Aesar™ 42023AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42023AP

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

37811-72-6
MFCD00048424
Isobutyl dodecanoate
256.43
Isobutyl laurate

Specifications

500mL
0.86g/mL
256.43
Isobutyl laurate
Isobutyl dodecanoate
37811-72-6
Liquid
MFCD00048424
150° (302°F)
≥99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit