Alfa Aesar™ Isobutyl dodecanoate, 99+%

Manufacturer: Alfa Aesar™ 42023AC

Quantity 25mL

 View more versions of this product

Catalog No. AA42023AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 37811-72-6
    Formula Weight 256.43
    MDL Number MFCD00048424
    Synonym Isobutyl laurate
    Chemical Name or Material Isobutyl dodecanoate

    Specifications

    Formula Weight 256.43
    Density 0.86g/mL
    MDL Number MFCD00048424
    Synonym Isobutyl laurate
    Physical Form Liquid
    CAS 37811-72-6
    Flash Point 150° (302°F)
    Quantity 25mL
    Chemical Name or Material Isobutyl dodecanoate
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit