Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isobutyl isothiocyanate, 97%

Click to view available options
Quantity:
5g
25g
100g
591-82-2
C5H9NS
115.194
MFCD00022062
NSDDRJXKROCWRZ-UHFFFAOYSA-N
isobutyl isothiocyanate,i-butyl isothiocyanate,isobutylisothiocyanate,2-methylpropyl isothiocyanate,isothiocyanic acid, isobutyl ester,1-isothiocyanato-2-methyl-propane,unii-a498zc6z4f,isothiocyanic acid isobutyl ester,propane, 1-isothiocyanato-2-methyl,propane, 1-isothiocyanato-2-methyl-9ci
68960
1-isothiocyanato-2-methylpropane
CC(C)CN=C=S
591-82-2
C5H9NS
115.194
MFCD00022062
NSDDRJXKROCWRZ-UHFFFAOYSA-N
isobutyl isothiocyanate,i-butyl isothiocyanate,isobutylisothiocyanate,2-methylpropyl isothiocyanate,isothiocyanic acid, isobutyl ester,1-isothiocyanato-2-methyl-propane,unii-a498zc6z4f,isothiocyanic acid isobutyl ester,propane, 1-isothiocyanato-2-methyl,propane, 1-isothiocyanato-2-methyl-9ci
68960
1-isothiocyanato-2-methylpropane
CC(C)CN=C=S
Applications
It is isothiocyanate compound used for proteomics research.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Refractive Index 1.498
Sensitivity Moisture sensitive
Density 0.94
Flash Point 24°C (75°F)
Boiling Point 160°C to 162°C
Odor Pungent
Quantity 100g
UN Number UN2924
Beilstein 1740371
Solubility Information Hydrolyzes with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material Isobutyl isothiocyanate
Show More Show Less

GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 209-733-2

RTECSNumber : NX8780100

TSCA : No

Recommended Storage : Ambient temperatures