Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutylene oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
558-30-5
C4H8O
72.11
MFCD00066354
GELKGHVAFRCJNA-UHFFFAOYSA-N
isobutylene oxide,1,2-epoxy-2-methylpropane,isobutene oxide,isobutylene epoxide,isobutyleneoxide,oxirane, 2,2-dimethyl,1,2-epoxyisobutane,1,1-dimethylethylene oxide,1,2-isobutylene oxide,2-methyl-1-propene oxide
11208
2,2-dimethyloxirane
CC1(CO1)C
558-30-5
C4H8O
72.11
MFCD00066354
GELKGHVAFRCJNA-UHFFFAOYSA-N
isobutylene oxide,1,2-epoxy-2-methylpropane,isobutene oxide,isobutylene epoxide,isobutyleneoxide,oxirane, 2,2-dimethyl,1,2-epoxyisobutane,1,1-dimethylethylene oxide,1,2-isobutylene oxide,2-methyl-1-propene oxide
11208
2,2-dimethyloxirane
CC1(CO1)C
Viscosity 0.513 mm2/s (20°C)
Specific Gravity 0.808
Packaging Glass bottle
Refractive Index 1.3710 to 1.3730
Quantity 25mL
Density 0.8080g/mL
Infrared Spectrum Authentic
Flash Point −32°C
Boiling Point 51.0°C to 52.0°C
Color Colorless
Assay Percent Range 98%
Beilstein 17, V,1, 58
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether
Formula Weight 72.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Isobutylene oxide, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-193-8

RTECSNumber : TZ3580000

TSCA : TSCA