Learn More
Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, ACROS Organics™
Specifications
Quantity | 100mL |
Density | 0.8300g/mL |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.83 |
Flash Point | −040°C |
Appearance | Grey to yellow-brown solution |
Linear Formula | (CH3)2CHCH2MgCl |
Solubility Information | Solubility in water: reacts. |
Formula Weight | 116.87 |
Chemical Name or Material | Isobutylmagnesium chloride, 2M solution in diethyl ether |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger