Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.8300g/mL |
Flash Point | −040°C |
Packaging | AcroSeal™ Glass Bottle |
Molecular Formula | C4H9ClMg |
Linear Formula | (CH3)2CHCH2MgCl |
Quantity | 800 mL |
Specific Gravity | 0.83 |
Synonym | isobutylmagnesium chloride, magnesium, chloro 2-methylpropyl, isobutylmagnesium chloride solution, 2.0 m in diethyl ether, isobutylmagnesiumchloride, ibumgcl, i-bumgcl, i-butylmagnesium chloride, isobutyl magnesium chloride, chloro 2-methylpropyl magnesium, chloro-2-methylpropyl magnesium |
Solubility Information | Solubility in water: reacts. |
InChI Key | YUPZOKFCNKSJTR-UHFFFAOYSA-M |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction