Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isobutyramide, 99%

Click to view available options
Quantity:
25g
100g
500g
563-83-7
C4H9NO
87.122
MFCD00008019
WFKAJVHLWXSISD-UHFFFAOYSA-N
isobutyramide,propanamide, 2-methyl,2-methylpropionamide,isobutyrimidic acid,isobutylamide,isopropylformamide,dimethylacetoamide,unii-82uoe7b38z,dimethylacetamid,c-isopropylformamide
68424
2-methylpropanamide
CC(C)C(=O)N
563-83-7
C4H9NO
87.122
MFCD00008019
WFKAJVHLWXSISD-UHFFFAOYSA-N
isobutyramide,propanamide, 2-methyl,2-methylpropionamide,isobutyrimidic acid,isobutylamide,isopropylformamide,dimethylacetoamide,unii-82uoe7b38z,dimethylacetamid,c-isopropylformamide
68424
2-methylpropanamide
CC(C)C(=O)N
Applications
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

Solubility
Soluble in water.

Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.
Quantity 500g
Melting Point 127°C to 129°C
Boiling Point 216°C to 220°C
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material Isobutyramide

RTECSNumber : TX1475000

TSCA : TSCA