Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyric Acid, 99+%, Thermo Scientific Chemicals

Catalog Number AC122520250
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
79-31-2
C4H8O2
88.11
MFCD00002658
KQNPFQTWMSNSAP-UHFFFAOYSA-N
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
6590
CHEBI:16135
2-methylpropanoic acid
CC(C)C(O)=O
79-31-2
C4H8O2
88.11
MFCD00002658
KQNPFQTWMSNSAP-UHFFFAOYSA-N
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
6590
CHEBI:16135
2-methylpropanoic acid
CC(C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
Melting Point -47.0°C
Boiling Point 153.0°C to 154.0°C
Color Colorless
Assay Percent Range 99+%
Linear Formula (CH3)2CHCO2H
Beilstein 02, 288
Merck Index 15, 5200
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: miscible with alcohol, chloroform and ether
Formula Weight 88.11
Percent Purity ≥99%
Density 0.9500g/mL
Quantity 25 g
Flash Point 59°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3920 to 1.3940
Specific Gravity 0.95
Chemical Name or Material Isobutyric acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-195-7
RTECSNumber NQ4375000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit