Learn More
Isobutyryl chloride, 98%, Thermo Scientific Chemicals
Description
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.
Solubility
Miscible with chloroform, glacial acetic acid, ether, toluene, methylene chloride and benzene.
Notes
Moisture sensitive. Incompatible with bases, water, strong oxidizing agents and alcohols.
Specifications
Melting Point | ∽-90°C |
Density | 1.017 |
Boiling Point | 91°C to 93°C |
Flash Point | 8°C (46°F) |
Assay Percent Range | 0.98 |
Refractive Index | 1.407 |
Quantity | 100 g |
UN Number | UN2395 |
Beilstein | 605393 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H225-H290-H302-H314-H330 |
Hazard Statement | GHS H Statement H225-H314 |
Precautionary Statement | P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRYL CHLORIDE |
EINECSNumber | 201-194-1 |
RTECSNumber | UC3944000 |
TSCA | Yes |
storageNote1 | Moisture Sensitive |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.