Learn More
Cayman ChemicalSupplier Diversity Partner Isobutyryl-L-carnitine, 25518-49-4, 10 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 1744010
A natural 4-carbon acylcarnitine that is involved in fatty acid oxidation and organic acid metabolism
Catalog No. NC1474068