Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isochroman 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
493-05-0
C9H10O
134.18
MFCD00006913
HEBMCVBCEDMUOF-UHFFFAOYSA-N
isochroman,isochromane,3,4-dihydro-1h-2-benzopyran,1h-2-benzopyran, 3,4-dihydro,3,4-dihydro-1h-benzo c pyran,pubchem20553,acmc-209kfu,isochroman 10g,ksc237m1l
96266
CHEBI:33225
3,4-dihydro-1H-isochromene
C1COCC2=CC=CC=C21
493-05-0
C9H10O
134.18
MFCD00006913
HEBMCVBCEDMUOF-UHFFFAOYSA-N
isochroman,isochromane,3,4-dihydro-1h-2-benzopyran,1h-2-benzopyran, 3,4-dihydro,3,4-dihydro-1h-benzo c pyran,pubchem20553,acmc-209kfu,isochroman 10g,ksc237m1l
96266
CHEBI:33225
3,4-dihydro-1H-isochromene
C1COCC2=CC=CC=C21
Boiling Point 75°C (4.0 mbar)
Flash Point 65°C
Specific Gravity 1.06
Refractive Index 1.5430 to 1.546
Density 1.0600g/mL
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Beilstein 17, 53
Formula Weight 134.18
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Isochroman, 99%
Show More Show Less

EINECSNumber : 207-774-0