Learn More
BioVision IncSupplier Diversity Partner Isocitrate Dehydrogenase Activity Colorimetric Assay Kit: Colorimetric Assay for Measuring IDH Activity in Serum, Tissues, Cells etc. Simple, Sensitive and Reliable. Detection Limit: 0.01 mU. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K756100
It is commonly measured clinically as a part of a diagnostic liver function test to determine liver health. Diagnostically, it is almost always measured in units/liter (U/L). In BioVision’s ALT Assay Kit, ALT catalyzes the transfer of an amino group from alanine to ⍺-ketoglutarate, the products of this reversible transamination reaction being pyruvate and glutamate. The pyruvate is detected in a reaction that concomitantly converts a nearly colorless probe to both color (λmax = 570 nm) and fluorescence (Ex/Em = 535/587 nm). The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput activity assay of ALT with a detection limit of 10 mU per well. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9880