Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isohexane, for pesticide residue analysis, contains <5% n-Hexane, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass Bottle
73513-42-5
C6H14
86.18
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
73513-42-5
C6H14
86.18
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
For residue analysis
Packaging Glass Bottle
Density 0.6600g/mL
Boiling Point 60.0°C
Specific Gravity 0.66
Residue after Evaporation 2ppm max.
Water 0.01% max. (coulometric)
Color Undesignated
Flash Point −23°C
Melting Point -154.0°C
Refractive Index 1.3700 to 1.3720
Quantity 2.5L
Linear Formula CH3CH2CH2CH(CH3)2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 86.18
Physical Form Liquid
Grade For pesticide residue analysis
Chemical Name or Material Isohexane, for residue and pesticides analysis, >5% n-Hexane
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger