Learn More
Mystaire™ Isola™ Filtered Workstation HEPA Filters
For use with Isola™ filtered workstations. Provides operator safety from potentially harmful airborne particulate.
$374.00 - $389.00
Specifications
Material High Efficiency Particulate Air
Product Type HEPA Filter
Filter Type Particulate
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
15-338-950
View Documents
Mystaire™
MY4020
36 and 72 in. wide Isola™ Filtered Workstation
Each for $374.00
Add to cart
15-338-951
View Documents
Mystaire™
MY4120
48 in. wide Isola™ Filtered Workstation
Each for $389.00
Add to cart
Description
Description
  • Provides 99.97% efficient down to 0.3 micron particles
Specifications
Specifications
High Efficiency Particulate Air
HEPA Filter
Particulate