Learn More
Biological Specialty CorpSupplier Diversity Partner Isolated Human PBMC's, 1 billion cells, fresh DFS Item
Manufacturer:  Biological Specialty CorpSupplier Diversity Partner 2140010
Mononuclear cells are isolated from whole blood or a leukopack using ficoll. The red blood cells are lysed. The MNC’s are washed and shipped according to your specifications. Minimum Cell Count: 370 million cells
Catalog No. NC0075382