Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoniazid, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

IS111, 54-85-3, C6H7N3O

Spectrum™ Isoniazid, USP is an organic compound that is also known as isonicotinylhydrazine. It is a medication used to treat and prevent tuberculosis. It is considered generally inexpensive and easily tolerated. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Melting Range 170-173°C
Loss on Drying 0.01
Quantity 25 kg
CAS 54-85-3
CAS Max % 1
Molecular Formula C6H7N3O
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
SMILES NNC(=O)C1=CC=NC=C1
IUPAC Name pyridine-4-carbohydrazide
Molecular Weight (g/mol) 137.14
Percent Purity 98 to 102%
Show More Show Less