Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinamide, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1453-82-3
C6H6N2O
122.127
MFCD00006432
VFQXVTODMYMSMJ-UHFFFAOYSA-N
isonicotinamide,4-pyridinecarboxamide,4-carbamoylpyridine,isonicotinic acid amide,isonicotineamide,gamma-pyridinecarboxamide,pyridine-4-carboxylic acid amide,unii-4h3bh6yx9q,.gamma.-pyridinecarboxamide,4h3bh6yx9q
15074
CHEBI:6031
pyridine-4-carboxamide
C1=CN=CC=C1C(=O)N
1453-82-3
C6H6N2O
122.127
MFCD00006432
VFQXVTODMYMSMJ-UHFFFAOYSA-N
isonicotinamide,4-pyridinecarboxamide,4-carbamoylpyridine,isonicotinic acid amide,isonicotineamide,gamma-pyridinecarboxamide,pyridine-4-carboxylic acid amide,unii-4h3bh6yx9q,.gamma.-pyridinecarboxamide,4h3bh6yx9q
15074
CHEBI:6031
pyridine-4-carboxamide
C1=CN=CC=C1C(=O)N
Quantity 100g
Color White
Infrared Spectrum Authentic
Melting Point 155°C to 158°C
Assay Percent Range 99%
Beilstein 22, 46
Formula Weight 122.13
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Isonicotinamide

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

Warning