Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isonicotinic acid, 99%

Click to view available options
Quantity:
100g
500g
2500g
55-22-1
C6H5NO2
123.111
MFCD00006429
TWBYWOBDOCUKOW-UHFFFAOYSA-N
isonicotinic acid,4-pyridinecarboxylic acid,4-picolinic acid,4-carboxypyridine,p-pyridinecarboxylic acid,gamma-picolinic acid,gamma-pyridinecarboxylic acid,acide iso-nicotinique,1,4-dihydroisonicotinic acid,acide iso-nicotinique french
5922
CHEBI:6032
pyridine-4-carboxylic acid
C1=CN=CC=C1C(=O)O
55-22-1
C6H5NO2
123.111
MFCD00006429
TWBYWOBDOCUKOW-UHFFFAOYSA-N
isonicotinic acid,4-pyridinecarboxylic acid,4-picolinic acid,4-carboxypyridine,p-pyridinecarboxylic acid,gamma-picolinic acid,gamma-pyridinecarboxylic acid,acide iso-nicotinique,1,4-dihydroisonicotinic acid,acide iso-nicotinique french
5922
CHEBI:6032
pyridine-4-carboxylic acid
C1=CN=CC=C1C(=O)O
Applications
It acts as an useful isomer of nicotinic acid.

Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.

Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point ~310°C (sublimation)
Boiling Point 260°C (15mmHg)
Quantity 500g
pH 3.0 to 4.0
Beilstein 109599
Merck Index 14,5187
Solubility Information Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Isonicotinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-228-2

RTECSNumber : NS1103000

TSCA : Yes

Recommended Storage : Ambient temperatures