Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinic Acid Hydrazide 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
54-85-3
C6H7N3O
137.14
MFCD00006426
QRXWMOHMRWLFEY-UHFFFAOYSA-N
isoniazid,isonicotinic acid hydrazide,isonicotinohydrazide,rimifon,isoniazide,nydrazid,isonicotinic hydrazide,laniazid,isonicotinylhydrazine,andrazide
3767
CHEBI:6030
pyridine-4-carbohydrazide
C1=CN=CC=C1C(=O)NN
54-85-3
C6H7N3O
137.14
MFCD00006426
QRXWMOHMRWLFEY-UHFFFAOYSA-N
isoniazid,isonicotinic acid hydrazide,isonicotinohydrazide,rimifon,isoniazide,nydrazid,isonicotinic hydrazide,laniazid,isonicotinylhydrazine,andrazide
3767
CHEBI:6030
pyridine-4-carbohydrazide
C1=CN=CC=C1C(=O)NN
Melting Point 171°C to 173°C
Infrared Spectrum Authentic
Quantity 100g
Packaging Plastic bottle
Color Colorless or White
Loss on Drying 0.5% max. (1 g, 105°C)
Assay Percent Range 99%
Beilstein 22, I, 504
Merck Index 15, 5232
Solubility Information Solubility in water: 14% in water (25 c). Other solubilities: 2% in alcohol (25 c),0.1% in chloroform
Formula Weight 137.14
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98.5% (on dried substance)
Chemical Name or Material Isonicotinic acid hydrazide, 99%
Show More Show Less

GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 200-214-6

RTECSNumber : NS1751850

TSCA : TSCA