Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinoyl chloride hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
Packaging:
Plastic bottle
39178-35-3
C6H4ClNO·ClH
178.02
MFCD00012830
BNTRVUUJBGBGLZ-UHFFFAOYSA-N
isonicotinoyl chloride hydrochloride,pyridine-4-carbonyl chloride hydrochloride,isonicotinoylchloride hydrochloride,4-pyridinecarbonyl chloride hydrochloride,4-pyridinecarbonyl chloride, hydrochloride,isonicotinoylchloridehcl,pubchem23974,acmc-1bn6p,ksc223m5d,4-pyridinecarbonyl chloride-hcl
12262826
pyridine-4-carbonyl chloride;hydrochloride
C1=CN=CC=C1C(=O)Cl.Cl
39178-35-3
C6H4ClNO·ClH
178.02
MFCD00012830
BNTRVUUJBGBGLZ-UHFFFAOYSA-N
isonicotinoyl chloride hydrochloride,pyridine-4-carbonyl chloride hydrochloride,isonicotinoylchloride hydrochloride,4-pyridinecarbonyl chloride hydrochloride,4-pyridinecarbonyl chloride, hydrochloride,isonicotinoylchloridehcl,pubchem23974,acmc-1bn6p,ksc223m5d,4-pyridinecarbonyl chloride-hcl
12262826
pyridine-4-carbonyl chloride;hydrochloride
C1=CN=CC=C1C(=O)Cl.Cl
Color Beige to White
Packaging Plastic bottle
Melting Point 153.0°C to 161.0°C
Quantity 250g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Beilstein 22, 46
Formula Weight 178.02
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Isonicotinoyl chloride hydrochloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 254-331-2