Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isonipecotamide, 98%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
Specifications
Melting Point | 146°C to 150°C |
Loss on Drying | 2% max. (70°C) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Color | White to Yellow |
Water | 2% max. (K.F.) |
Quantity | 25g |
Sulfated Ash | 0.2% max. |
Assay Percent Range | 98% |
Solubility Information | Solubility in water: soluble |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 254-501-6